Assay Details
| Gene Symbol: DACH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260046.2 | XP_008258268.1 | 10 - 11 | 2724 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DACH1| Gene Symbol: DACH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260046.2 | XP_008258268.1 | 10 - 11 | 2724 | 79 | |
Set Membership: |
|