Assay Details
| Gene Symbol: CCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719406.2 | XP_002719452.1 | 2 | 309 | 65 | |
| GenBank mRNA | EU348829.1 | - | 1 | 151 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCR10| Gene Symbol: CCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719406.2 | XP_002719452.1 | 2 | 309 | 65 | |
| GenBank mRNA | EU348829.1 | - | 1 | 151 | 65 | |
Set Membership: |
|