Assay Details
| Gene Symbol: G6PC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719415.3 | XP_002719461.2 | 3 - 4 | 663 | 101 | |
| GenBank mRNA | EU520488.1 | - | 2 - 3 | 192 | 101 | |
More Information
Set Membership: |
|

G6PC
Genomic Map