Assay Details
| Gene Symbol: IL18R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164863.1 | NP_001158335.1 | 7 - 8 | 952 | 66 | |
| XM_008252716.2 | XP_008250938.1 | 8 - 9 | 1501 | 66 | ||
| GenBank mRNA | EU731009.1 | - | 7 - 8 | 952 | 66 | |
More Information
Set Membership: |
|

IL18R1
Genomic Map