Assay Details
| Gene Symbol: DHFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261878.2 | XP_008260100.2 | 4 - 5 | 72 | ||
| GenBank mRNA | FJ687582.1 | - | 1 | 446 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DHFR| Gene Symbol: DHFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261878.2 | XP_008260100.2 | 4 - 5 | 72 | ||
| GenBank mRNA | FJ687582.1 | - | 1 | 446 | 72 | |
Set Membership: |
|