Assay Details
| Gene Symbol: GCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718258.3 | XP_002718304.1 | 5 - 6 | 771 | 66 | |
| GenBank mRNA | FJ687583.1 | - | 5 - 6 | 452 | 66 | |
More Information
Set Membership: |
|

GCH1
Genomic Map