Assay Details
| Gene Symbol: UGT2C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195805.1 | NP_001182734.1 | 2 - 3 | 870 | 89 | |
| GenBank mRNA | L01083.1 | - | 2 - 3 | 789 | 89 | |
More Information
Set Membership: |
|

UGT2C1
Genomic Map