Assay Details
| Gene Symbol: CA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017341428.1 | XP_017196917.1 | 806 | 71 | ||
| GenBank mRNA | M10412.1 | - | 368 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017341428.1 | XP_017196917.1 | 806 | 71 | ||
| GenBank mRNA | M10412.1 | - | 368 | 71 | ||
Set Membership: |
|