Assay Details
| Gene Symbol: CSNK2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160283.1 | NP_001153755.1 | 1 - 2 | 113 | 93 | |
| GenBank mRNA | M98450.1 | - | 1 - 2 | 113 | 93 | |
| S56242.1 | - | 1 - 2 | 113 | 93 | ||
More Information
Set Membership: |
|

CSNK2B
Genomic Map