Assay Details
| Gene Symbol: KCNA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082676.2 | NP_001076145.2 | 1 | 2037 | 70 | |
| GenBank mRNA | U38182.1 | - | 1 | 2046 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNA10| Gene Symbol: KCNA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082676.2 | NP_001076145.2 | 1 | 2037 | 70 | |
| GenBank mRNA | U38182.1 | - | 1 | 2046 | 70 | |
Set Membership: |
|