Assay Details
| Gene Symbol: IGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082026.1 | NP_001075495.1 | 1 - 2 | 177 | 68 | |
| XM_008256716.2 | XP_008254938.1 | 2 - 3 | 513 | 68 | ||
| XM_008256717.2 | XP_008254939.1 | 2 - 3 | 280 | 68 | ||
| XM_008256718.2 | XP_008254940.1 | 2 - 3 | 506 | 68 | ||
| XM_008256719.2 | XP_008254941.1 | 2 - 3 | 498 | 68 | ||
| XM_008256720.2 | XP_008254942.1 | 2 - 3 | 267 | 68 | ||
| XM_017341617.1 | XP_017197106.1 | 2 - 3 | 261 | 68 | ||
| GenBank mRNA | AF022961.1 | - | 1 - 2 | 177 | 68 | |
| U75390.1 | - | 1 - 2 | 177 | 68 | ||
More Information
Set Membership: |
|

IGF1
Genomic Map