Assay Details
| Gene Symbol: PPP1CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101706.1 | NP_001095176.1 | 706 | 78 | ||
| GenBank mRNA | X07798.1 | - | 2 - 3 | 559 | 78 | |
| X14832.1 | - | 563 | 78 | |||
| Y00701.1 | - | 706 | 78 | |||
More Information
Set Membership: |
|

PPP1CA
Genomic Map