Assay Details
| Gene Symbol: LPAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264963.2 | XP_008263185.1 | 2 | 501 | 78 | |
| GenBank mRNA | AF404276.1 | - | 1 | 245 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LPAR3| Gene Symbol: LPAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264963.2 | XP_008263185.1 | 2 | 501 | 78 | |
| GenBank mRNA | AF404276.1 | - | 1 | 245 | 78 | |
Set Membership: |
|