Assay Details
| Gene Symbol: RACK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262008.2 | XP_008260230.1 | 1 - 2 | 228 | 72 | |
| GenBank mRNA | AY249909.1 | - | 1 - 2 | 102 | 72 | |
More Information
Set Membership: |
|

RACK1
Genomic Map