Assay Details
| Gene Symbol: CD247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082002.1 | NP_001075471.1 | 1 - 2 | 101 | 63 | |
| XM_008263943.2 | XP_008262165.1 | 1 - 2 | 63 | |||
| GenBank mRNA | AB035152.1 | - | 1 - 2 | 101 | 63 | |
More Information
Set Membership: |
|

CD247
Genomic Map