Assay Details
| Gene Symbol: CHST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269989.2 | XP_008268211.1 | 3 | 956 | 75 | |
| XM_008269990.2 | XP_008268212.1 | 3 | 1504 | 75 | ||
| XM_008269991.2 | XP_008268213.1 | 3 | 1091 | 75 | ||
| GenBank mRNA | AB073106.1 | - | 1 | 144 | 75 | |
More Information
Set Membership: |
|

CHST3
Genomic Map