Assay Details
| Gene Symbol: IRAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017345354.1 | XP_017200843.1 | 16 - 17 | 60 | ||
| XM_017345357.1 | XP_017200846.1 | 17 - 18 | 60 | |||
| GenBank mRNA | AY587767.1 | - | 3 - 4 | 234 | 60 | |
More Information
Set Membership: |
|

IRAG1
Genomic Map