Assay Details
| Gene Symbol: SOX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271763.2 | XP_008269985.1 | 1 - 2 | 824 | 61 | |
| GenBank mRNA | AY598935.1 | - | 1 - 2 | 425 | 61 | |
More Information
Set Membership: |
|

SOX9
Genomic Map