Assay Details
| Gene Symbol: GJA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198948.1 | NP_001185877.1 | 1 | 236 | 84 | |
| XM_008263297.2 | XP_008261519.1 | 2 | 459 | 84 | ||
| GenBank mRNA | AY576039.1 | - | 1 | 185 | 84 | |
| DQ242486.1 | - | 1 | 53 | 84 | ||
More Information
Set Membership: |
|

GJA1
Genomic Map