Assay Details
| Gene Symbol: KCNJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082198.1 | NP_001075667.1 | 3 | 1507 | 80 | |
| XM_017348806.1 | XP_017204295.1 | 2 | 1529 | 80 | ||
| XM_017348807.1 | XP_017204296.1 | 2 | 1598 | 80 | ||
| GenBank mRNA | AF021138.1 | - | 1 | 1148 | 80 | |
| D21057.1 | - | 3 | 1507 | 80 | ||
More Information
Set Membership: |
|

KCNJ2
Genomic Map