Assay Details
| Gene Symbol: TIMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195682.1 | NP_001182611.1 | 4 - 5 | 438 | 70 | |
| GenBank mRNA | AF069714.1 | - | 4 - 5 | 335 | 70 | |
| EF472914.1 | - | 4 - 5 | 337 | 70 | ||
More Information
Set Membership: |
|

TIMP3
Genomic Map