Assay Details
| Gene Symbol: SNURF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082714.1 | NP_001076183.1 | 1 | 63 | 68 | |
| GenBank mRNA | AF101043.1 | - | 1 | 63 | 68 | |
| Gene Symbol: LOC108175375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344764.1 | XP_017200253.1 | 1 | 42 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001082714.1, XM_017344764.1 transcripts.
Set Membership: |
|

SNURF
Genomic Map