Assay Details
| Gene Symbol: FABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710655.3 | XP_002710701.1 | 1 - 2 | 142 | 74 | |
| GenBank mRNA | AF136241.1 | - | 1 - 2 | 49 | 74 | |
More Information
Set Membership: |
|

FABP4
Genomic Map