Assay Details
| Gene Symbol: LOC100009353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344649.1 | XP_017200138.1 | 1 | 144 | 72 | |
| GenBank mRNA | AF202899.1 | - | 1 | 144 | 72 | |
More Information
Set Membership: |
|

LOC100009353
Genomic Map