Assay Details
Gene Symbol: LOC100144340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716358.3 | XP_002716404.1 | 1 | 333 | 63 | |
GenBank mRNA | EF222473.1 | - | 2 - 3 | 145 | 63 |
Gene Symbol: LOC100340199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002707914.3 | XP_002707960.1 | 1 | 328 | 63 |
Gene Symbol: LOC100346276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712764.3 | XP_002712810.1 | 1 | 323 | 63 |
Gene Symbol: LOC100354806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717506.3 | XP_002717552.1 | 1 | 327 | 63 |
Gene Symbol: RPSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713098.3 | XP_002713144.1 | 3 - 4 | 423 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
