Assay Details
| Gene Symbol: ADRA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082380.1 | NP_001075849.1 | 2 | 79 | ||
| GenBank mRNA | AF157505.1 | - | 2 | 79 | ||
| AF157506.1 | - | 2 | 79 | |||
| S75999.1 | - | 2 | 344 | 79 | ||
| U81982.1 | - | 2 | 79 | |||
More Information
Set Membership: |
|

ADRA1A
Genomic Map