Assay Details
| Gene Symbol: LOC100009480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082723.1 | NP_001076192.1 | 69 | 72 | ||
| GenBank mRNA | AF308620.1 | - | 69 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100009480| Gene Symbol: LOC100009480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082723.1 | NP_001076192.1 | 69 | 72 | ||
| GenBank mRNA | AF308620.1 | - | 69 | 72 | ||
Set Membership: |
|