Assay Details
| Gene Symbol: SULT2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081987.1 | NP_001075456.1 | 348 | 79 | ||
| XM_017339932.1 | XP_017195421.1 | 425 | 79 | |||
| GenBank mRNA | AB006053.1 | - | 348 | 79 | ||
More Information
Set Membership: |
|

SULT2A1
Genomic Map