Assay Details
| Gene Symbol: SULT2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081987.1 | NP_001075456.1 | 565 | 80 | ||
| GenBank mRNA | AB006053.1 | - | 565 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SULT2A1| Gene Symbol: SULT2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081987.1 | NP_001075456.1 | 565 | 80 | ||
| GenBank mRNA | AB006053.1 | - | 565 | 80 | ||
Set Membership: |
|