Assay Details
| Gene Symbol: CYP19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170921.2 | NP_001164392.1 | 2 - 3 | 413 | 68 | |
| GenBank mRNA | Z68271.1 | - | 2 - 3 | 414 | 68 | |
| Z70301.1 | - | 2 - 3 | 414 | 68 | ||
| Z70302.1 | - | 2 - 3 | 414 | 68 | ||
More Information
Set Membership: |
|

CYP19A1
Genomic Map