Assay Details
| Gene Symbol: H2AZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170941.1 | NP_001164412.1 | 2 - 3 | 224 | 76 | |
| GenBank mRNA | AF030235.1 | - | 2 - 3 | 224 | 76 | |
More Information
Set Membership: |
|

H2AZ2
Genomic Map