Assay Details
| Gene Symbol: UGT2B16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171016.2 | NP_001164487.2 | 4 - 5 | 1102 | 111 | |
| XM_017347249.1 | XP_017202738.1 | 4 - 5 | 1135 | 111 | ||
| GenBank mRNA | U72742.1 | - | 4 - 5 | 111 | ||
More Information
Set Membership: |
|

UGT2B16
Genomic Map