Assay Details
| Gene Symbol: FSCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171280.1 | NP_001164751.1 | 4 - 5 | 1124 | 72 | |
| XM_008258187.2 | XP_008256409.1 | 4 - 5 | 1280 | 72 | ||
More Information
Set Membership: |
|

FSCN3
Genomic Map