Assay Details
| Gene Symbol: SERPINB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171283.1 | NP_001164754.1 | 6 - 7 | 766 | 88 | |
| XM_008261382.2 | XP_008259604.1 | 7 - 8 | 88 | |||
| XM_008261383.2 | XP_008259605.1 | 7 - 8 | 88 | |||
| XM_008261384.2 | XP_008259606.1 | 6 - 7 | 88 | |||
More Information
Set Membership: |
|

SERPINB13
Genomic Map