Assay Details
| Gene Symbol: SCNM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171298.1 | NP_001164769.1 | 4 - 5 | 306 | 59 | |
| XM_008264020.2 | XP_008262242.1 | 4 - 5 | 274 | 59 | ||
| XM_008264021.2 | XP_008262243.1 | 3 - 4 | 434 | 59 | ||
More Information
Set Membership: |
|

SCNM1
Genomic Map