Assay Details
| Gene Symbol: CKMT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171313.1 | NP_001164784.1 | 5 - 6 | 754 | 62 | |
| XM_008268820.2 | XP_008267042.1 | 6 - 7 | 1001 | 62 | ||
| XM_008268821.2 | XP_008267043.1 | 6 - 7 | 1012 | 62 | ||
| XM_017347911.1 | XP_017203400.1 | 6 - 7 | 953 | 62 | ||
| XM_017347912.1 | XP_017203401.1 | 6 - 7 | 878 | 62 | ||
More Information
Set Membership: |
|

CKMT1B
Genomic Map