Assay Details
Gene Symbol: RBM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171333.1 | NP_001164804.1 | 1 | 1443 | 86 |
Gene Symbol: CPNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008255973.2 | XP_008254195.1 | 3 | 1683 | 86 | |
XM_008255974.2 | XP_008254196.1 | 3 | 1621 | 86 | ||
XM_008255975.2 | XP_008254197.1 | 3 | 1537 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
