Assay Details
| Gene Symbol: NFS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171334.1 | NP_001164805.1 | 10 - 11 | 1126 | 60 | |
| XM_008255966.2 | XP_008254188.1 | 9 - 10 | 1034 | 60 | ||
More Information
Set Membership: |
|

NFS1
Genomic Map