Assay Details
| Gene Symbol: SLC22A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171346.1 | NP_001164817.1 | 8 - 9 | 1445 | 66 | |
| XM_017340987.1 | XP_017196476.1 | 8 - 9 | 1783 | 66 | ||
More Information
Set Membership: |
|

SLC22A4
Genomic Map