Assay Details
| Gene Symbol: SLC5A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171422.1 | 1944 | 87 | |||
| XM_008274929.2 | XP_008273151.1 | 4378 | 87 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC5A3| Gene Symbol: SLC5A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171422.1 | 1944 | 87 | |||
| XM_008274929.2 | XP_008273151.1 | 4378 | 87 | |||
Set Membership: |
|