Assay Details
| Gene Symbol: RLA-A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190434.1 | NP_001177363.1 | 1 - 2 | 93 | 64 | |
| XM_008262632.2 | XP_008260854.1 | 1 - 2 | 162 | 64 | ||
| XM_017344790.1 | XP_017200279.1 | 1 - 2 | 229 | 64 | ||
| GenBank mRNA | K02442.1 | - | 1 - 2 | 93 | 64 | |
More Information
Set Membership: |
|

RLA-A3
Genomic Map