Assay Details
| Gene Symbol: LIPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195638.1 | NP_001182567.1 | 5 - 6 | 856 | 65 | |
| GenBank mRNA | EU871041.1 | - | 5 - 6 | 856 | 65 | |
More Information
Set Membership: |
|

LIPG
Genomic Map