Assay Details
| Gene Symbol: FGF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195837.1 | NP_001182766.1 | 2 - 3 | 385 | 120 | |
| XM_008268784.2 | XP_008267006.1 | 3 - 4 | 929 | 120 | ||
More Information
Set Membership: |
|

FGF7
Genomic Map