Assay Details
| Gene Symbol: KCNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198937.1 | NP_001185866.1 | 153 | 70 | ||
| XM_008274920.2 | XP_008273142.1 | 395 | 70 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNE2| Gene Symbol: KCNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198937.1 | NP_001185866.1 | 153 | 70 | ||
| XM_008274920.2 | XP_008273142.1 | 395 | 70 | |||
Set Membership: |
|