Assay Details
| Gene Symbol: S1PR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198946.1 | NP_001185875.1 | 1 | 1050 | 64 | |
| XM_008263949.2 | XP_008262171.1 | 2 | 1556 | 64 | ||
| XM_008263950.2 | XP_008262172.1 | 2 | 1534 | 64 | ||
More Information
Set Membership: |
|

S1PR1
Genomic Map