Assay Details
| Gene Symbol: SELENOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199846.1 | NP_001186775.1 | 2 - 3 | 266 | 71 | |
| GenBank mRNA | CU465962.1 | - | 3 - 4 | 624 | 71 | |
| EC618869.1 | - | 2 - 3 | 306 | 71 | ||
More Information
Set Membership: |
|

SELENOP
Genomic Map