Assay Details
| Gene Symbol: EIF4EBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204114.1 | NP_001191043.1 | 2 - 3 | 351 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF4EBP3| Gene Symbol: EIF4EBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204114.1 | NP_001191043.1 | 2 - 3 | 351 | 66 | |
Set Membership: |
|