Assay Details
| Gene Symbol: NME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204749.1 | NP_001191678.1 | 1 - 2 | 128 | 59 | |
| XM_008271104.2 | XP_008269326.1 | 2 - 3 | 269 | 59 | ||
More Information
Set Membership: |
|

NME1
Genomic Map