Assay Details
| Gene Symbol: STON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204846.1 | NP_001191775.1 | 2 - 3 | 2124 | 71 | |
| XM_008252788.2 | XP_008251010.1 | 3 - 4 | 2254 | 71 | ||
| XM_008252794.2 | XP_008251016.1 | 3 - 4 | 2288 | 71 | ||
| XM_008252798.2 | XP_008251020.1 | 3 - 4 | 2253 | 71 | ||
More Information
Set Membership: |
|

STON1
Genomic Map