Assay Details
| Gene Symbol: UGT1-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089322.1 | NP_001082791.1 | 890 | 93 | ||
| GenBank mRNA | U09101.1 | - | 890 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UGT1-4| Gene Symbol: UGT1-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089322.1 | NP_001082791.1 | 890 | 93 | ||
| GenBank mRNA | U09101.1 | - | 890 | 93 | ||
Set Membership: |
|